Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYTL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112958
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112958 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112958 Supplier Invitrogen™ Supplier No. PA5112958
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SYTL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

May act as Rab effector protein and play a role in vesicle trafficking. Binds phospholipids in the presence of calcium ions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYTL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SYTL3
Gene Accession No. Q4VX76
Gene Alias A430092N21; Exophilin-6; mFLJ00307; RGD1561992; Slp3; Slp3-a; Slp3-b; synaptotagmin like 3; synaptotagmin-like 3; synaptotagmin-like protein 3; Sytl3
Gene Symbols SYTL3
Host Species Rabbit
Immunogen Recombinant Human Synaptotagmin-like protein 3 protein (1-300AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94120
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.