Learn More
Selleck Chemical LLC SZL P1-41-5mg
SDP
Supplier:  Selleck Chemical LLC S97245MG
Encompass_Preferred
SZL P1-41 (compound #25) is a specific inhibitor of S-phase kinase-associated protein 2 (Skp2) that indeed binds to Skp2, prevents Skp2-Skp1 interaction and inhibits Skp2 SCF E3 ligase activity, which consequently suppresses survival of cancer cells and cancer stem cells. SZL P1-41 causes higher apoptosis rates in cancer cells.
Catalog No. 50-238-9747
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.