Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ T 112
GREENER_CHOICE

Catalog No. p-7249490
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 76-901-0 Supplier Tocris Bioscience™ Supplier No. 7690/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Estrogen-related receptor γ agonist; promotes maturation of hiPSC-derived cardiomyocytes

T 112 is an estrogen-related receptor gamma (ERRγ) agonist. Promotes maturation of hiPSC-derived cardiomyocytes. T 112 accelerates metabolic and sarcomere maturation in cardiomyocytes, as well as maturation of contractile and electrophysiological properties and ultrastructure organization, when used alone or in combination with other small molecules.

Specifications

Chemical Name or Material 2-[2-(4-Bromophenyl)-1H-benzimidazol-6-yl]-2,3-dihydro-5-(4-morpholinyl)-1H-isoindol-1-one
CAS 2376876-87-6
Quantity 10 mg
Target Estrogen and Related Receptor Agonists
Molecular Formula C25H21BrN4O2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.