Learn More
AdipoGen T-2 Toxin
SDP
Supplier:  AdipoGen AGCN20473M001
Encompass_Preferred
Chemical. CAS 21259-20-1. Formula C24H34O9. MW 466.5. Isolated from fungus Fusarium tricinctum. Common trichothecene mycotoxin, which can infect grain crops causing alimentary toxic aleukia in humans and animals. Inhibits DNA and RNA synthesis in vivo and in vitro and can induce apoptosis. In vivo the compound rapidly metabolizes to HT-2 mycotoxin a major metabolite. Inhibits protein synthesis through its high binding affinity to peptidyl transferase, which is an integral part of the 60S ribosomal subunit, resulting in activation of JNK/p38 MAPKs. Interferes with the metabolism of membrane phospholipids and increases liver lipid peroxides. Increases blood-brain barrier permeability and inhibits monoamine oxidase activity in brain. Reactive oxygen species ROS inducer.
Catalog No. 50-256-2517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.