Learn More
Apexbio Technology LLC TAE226 (NVP-TAE226), 10mg. Cas: 761437-28-9 MFCD: MFCD12031516. FAK inhibitor,potent and ATP-competitive
SDP
Supplier:  Apexbio Technology LLC A855710
Encompass_Preferred
TAE226 (NVP-TAE226) is a selective inhibitor of FAK with IC50 value of 5.5 nM [1].Focal Adhesion Kinase (FAK) also known as PTK2 protein tyrosine kinase 2 (PTK2) is a focal adhesion-associated protein kinase and plays an important role in cellular adhesion and spreading processes [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5162
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.