Learn More
Invitrogen™ T-bet Monoclonal Antibody (eBio4B10 (4B10)), PE-Cyanine5, eBioscience™

Description
Description: The eBio4B10 monoclonal antibody reacts with mouse and human T-bet. T-bet is a Th1-specific T-box transcription factor critical to the development of the Th1 CD4+ T cell lineage. This is known based on the observations that T-Bet deficient mice have impaired Th1 cell development, and that ectopic expression of T-Bet results in development skewed to the Th1 lineage. T-Bet expression is induced by the Th1 cytokine IFN gamma, and T-Bet also regulates the expression of IFN gamma, likely, at least in part, through the modification of DNA accessibility and histone remodeling. In addition to IFN gamma, T-Bet is also known to regulate the expression of IL-12R beta and IL-2. Moreover, T-Bet plays a role in class-switch recombination in B-cells. Applications Reported: This eBio4B10 (4B10) antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBio4B10 (4B10) antibody has been tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to 'Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins' located at www.thermofisher.com/flowprotocols . This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in...
Specifications
Specifications
| Antigen | T-bet |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBio4B10 (4B10) |
| Concentration | 0.2 mg/mL |
| Conjugate | PE-Cyanine5 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Tbx21 |
| Gene Accession No. | Q9JKD8, Q9UL17 |
| Gene Alias | HGNC:11599; LOW QUALITY PROTEIN: T-box transcription factor TBX21; Tbet; T-bet; TBLYM; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor 21; T-box transcription factor TBX21; TBT1; TBX21; T-cell-specific T-box transcription factor T-bet; T-PET; transcription factor TBLYM; transcription factor T-box 21 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.