Learn More
Broadpharm t-Boc-N-amido-PEG2-amine, 153086-78-3, MFCD03788155, 1g
Supplier:  Broadpharm BP203011G
Encompass
Formula: C11H24N2O4, Purity: 98%, MW: 248.3, t-Boc-N-Amido-PEG2-amine is a PEG derivative containing an amino group and Boc-protected amino group. The hydrophilic PEG spacer increases solubility in aqueous media. The amino group is reactive with carboxylic acids, activated NHS esters, carbonyls (ketone, aldehyde) etc. The Boc group can be deprotected under mild acidic conditions to form the free amine.
Catalog No. NC1392679
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.