Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

t-Butyl Alcohol, 80% v/v (L-057), Reagents

Catalog No. 01335730
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-730 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-730 Supplier Reagents Holdings Llc Supplier No. CB18080010E2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

t-Butyl Alcohol, 80% v/v (L-057), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-65-0 , 7732-18-5
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Color Colorless
Quantity 10 L
Physical Form Liquid
Specific Gravity 0.8338
Percent Purity 80%
Grade Reagent
Chemical Name or Material t-Butyl Alcohol, 80% v/v (L-057)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.