Learn More
Bachem DOTA(OtBu)3 Hydrobromide
SDP
Supplier:  Bachem 4029066.0500
Encompass_Preferred
DOTA(OtBu)3 Hydrobromide, 4,7,10-Tri-(t-butyloxycarbonylmethyl)-1,4,7,10-tetraazacyclododecan-1-yl-acetic acid, 137076-54-1, C28H52N4O8, Mr 572.74, 500mg. t-Butyl ester-protected DOTA derivative allows the selective conjugation of the macrocyclic chelator to peptides during Fmoc-SPPS. DOTA forms highly stable chelates with radioactive isotopes of Ga,In,and Y, as well as with radionuclides of Lu, Gd, and other lanthanides.
Catalog No. 50-194-7402
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.