Learn More
Description
Specifications
Specifications
| Antigen | T Cell Exhaustion Marker (Flow) |
| Applications | CyTOF, Flow Cytometry |
| Conjugate | Unconjugated |
| Formulation | See individual datasheets |
| Gene Alias | exhaustion markers, immune cell exhaustion, immune checkpoint markers, inhibitory receptors, T cell exhaustion |
| Immunogen | See individual datasheets |
| Quantity | 9 Vials |
| Regulatory Status | RUO |
| Target Species | Human |
| Content And Storage | Storage is content dependent. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
