Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T Cell Exhaustion Marker (Flow-Directly Conjugated) Antibody Pack, Novus Biologicals™
SDP

Catalog No. NB176539 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
6 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176539 6 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176539 Supplier Novus Biologicals Supplier No. NBP312863
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antibody Pack

T Cell Exhaustion Marker (Flow-Directly Conjugated) Monoclonal specifically detects T Cell Exhaustion Marker (Flow-Directly Conjugated) in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen T Cell Exhaustion Marker (Flow-Directly Conjugated)
Applications Flow Cytometry
Classification Monoclonal
Conjugate Unconjugated
Formulation See individual datasheets
Gene Alias Immune cell exhaustion markers, immune checkpoint markers, inhibitory receptors, T cell exhaustion markers
Immunogen See individual datasheets
Quantity 6 Vials
Regulatory Status RUO
Target Species Human
Content And Storage Storage is content dependent.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.