Learn More
Sartorius T Cell Phenotyping 1x96
Supplier:  Sartorius 97099
Encompass
High-throughput: Designed for high-throughput immunophenotyping of T cells in 96- and 384-well plate format using 10L 96-well format and 5L 384-well format of cells.Biological insights: Generate functional data and option to analyze cell health from the same cell by multiplexing cell phenotypes and function in a single well.Flexibility: Choose additional cytokines to be added for further characterization of subpopulations.
Catalog No. 50-289-1737