Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty
Limited Lifetime Warranty
Specifications
Specifications
| Product Type | Hex Key T-handle |
| Material | Beryllium Copper |
| Size | 1/4 in. |
| Certifications/Compliance | Factory Mutual (FM) Approved |
Safety and Handling
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.