Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ T.T.C. Solution 5%

Catalog No. R663211
Change view
Click to view available options
Quantity:
1 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R663211 1 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R663211 Supplier Thermo Scientific™ Supplier No. SR0211C

This product is either new or inactive, and cannot be ordered online.

This item is not returnable. View return policy

Enumerate and identify coliform bacteria using this solution.

Oxoid TTC Solution is used for the enumeration and identification of coliform bacteria.

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Product Type Microbiology Media
Quantity 1 x 100 mL
Yield As required
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.