Learn More
Selleck Chemical LLC T0070907 S2871-25mg
SDP
Supplier:  Selleck Chemical LLC S287125MG
Encompass_Preferred
T0070907 is a potent inhibitor of PPAR (peroxisome proliferator activator receptor ) that induces G2/M arrest and enhances the effect of radiation in human cervical cancer cells through mitotic catastrophe It also acts as an antagonist of PPAR that suppresses breast cancer cell proliferation and motility via both PPARgamma-dependent and -independent mechanisms
Catalog No. 50-136-3114
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.