Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Testosterone Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco T0371ML
Description
A certified solution standard applicable to use in sports testing, urine drug testing, and diagnostic testing by LC-MS/MS or GC/MS. Testosterone, a hormone from the androgen group, is an important sex hormone and anabolic steroid often abused in sports.
Specifications
| 75-05-8, 58-22-0 | |
| MFCD00003654 | |
| 1.0 mg/mL in acetonitrile | |
| Ampule | |
| 2°C (35.6°F) - closed cup |
| C19H28O2 | |
| UN 1648 3 / PGII | |
| 288.42 | |
| 1 mL |
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction