Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Tadalafil Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0721ML

Catalog No. 01-002-069


Only null left
Add to Cart

Description

Description

Tadalafil is an erectile-dysfunction drug marketed under the trade name Cialis™. This Certified Spiking Solution™ is suitable for use in numerous testing applications such as clinical toxicology, forensic analysis, urine drug testing, or calibrator preparation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8, 171596-29-5
MFCD07771966
1.0 mg/mL in acetonitrile
Ampule
2°C (35.6°F) - closed cup
C22H19N3O4
UN 1648 3 / PGII
389.40
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

EINECSNumber : 200-835-2

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation