Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T16Ainh - A01, Tocris Bioscience™
SDP

Catalog No. 453810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-381-0 10 mg
45-385-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-381-0 Supplier R&D Systems Supplier No. 4538/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ca2+-activated Cl- channel transmembrane protein 16A (TMEM16A) inhibitor

Inhibitor of Ca2+-dependent Cl- channel (CaCC) transmembrane protein 16A (TMEM16A) (IC50 = 1.8 μM in A253 salivary gland epithelial cells). Inhibits EGF-induced increases in CaCC currents; blocks proliferation of tumor cells in vitro.

Chemical Identifiers

CAS 552309-42-9
Molecular Formula C19H20N4O3S2
Molecular Weight (g/mol) 416.514
InChI Key QSIYTNYMBWYHAA-UHFFFAOYSA-N
Synonym t16a inh-a01, t16ainh-a01, 2-5-ethyl-4-methyl-6-oxo-1,6-dihydropyrimidin-2-yl thio-n-4-4-methoxyphenyl thiazol-2-yl acetamide, 2-5-ethyl-1,6-dihydro-4-methyl-6-oxo-2-pyrimidinyl thio-n-4-4-methoxyphenyl-2-thiazolyl acetamide, 2-5-ethyl-4-hydroxy-6-methylpyrimidin-2-yl sulfanyl-n-4-4-methoxyphenyl-1,3-thiazol-2-yl acetamide, t16ainh-a hplc, 2-5-ethyl-4-hydroxy-6-methylpyrimidin-2-ylthio-n-4-4-methoxyphenyl thiazol-2-yl acetamide, 2-5-ethyl-4-methyl-6-oxo-3h-pyrimidin-2-yl sulfanyl-n-4-4-methoxyphenyl-1,3-thiazol-2-yl acetamide, 2-5-ethyl-6-methyl-4-oxo-1h-pyrimidin-2-yl sulfanyl-n-4-4-methoxyphenyl-1,3-thiazol-2-yl acetamide, n-4-4-methoxyphenyl thiazole-2-yl-2-4-hydroxy-5-ethyl-6-methylpyrimidine-2-ylthio acetamide
PubChem CID 3193184
IUPAC Name 2-[(5-ethyl-6-methyl-4-oxo-1H-pyrimidin-2-yl)sulfanyl]-N-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]acetamide
SMILES CCC1=C(NC(=NC1=O)SCC(=O)NC2=NC(=CS2)C3=CC=C(C=C3)OC)C

Specifications

Quantity 10 mg
Formula Weight 416.52
Percent Purity >98%
Chemical Name or Material T16Ainh - A01
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.