Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ T176B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112751
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112751 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112751 Supplier Invitrogen™ Supplier No. PA5112751
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

T176B Polyclonal Antibody for Western Blot, ICC/IF, ELISA

TMEM176B (also known as LR8) is a transmembrane protein belongs to the MS4A family of proteins. It may play a role in the process of maturation of dendritic cells. TMEM176B was first discovered in human lung fibroblasts and is associated with human small cell lung carcinoma. Studies suggest that TMEM176B might be involved in cancer and might serve as targets for antiangiogenic therapy of cancer patients. There are two isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen T176B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM176B
Gene Accession No. Q3YBM2
Gene Alias 1810009M01Rik; Clast1; LR8; LR8-like protein; MS4B2; Protein LR8; Tmem176b; Transmembrane protein 176B
Gene Symbols TMEM176B
Host Species Rabbit
Immunogen Recombinant Human Transmembrane protein 176B protein (1-64AA).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28959
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.