Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T3 DNA Ligase, 750000 U, G-Biosciences
SCI_ED

Catalog No. S001152
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001152 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001152 Supplier G-Biosciences Supplier No. 7862262
Add to Cart
Add to Cart

T3 DNA Ligase catalyzes the formation of a phosphodiester bond between a 5' phosphate and a 3' hydroxyl termini in duplex DNA; joining blunt and cohesive end termini as well as repairing single stranded nicks in duplex DNA.

  • T3 DNA Ligase displays a higher tolerance for salt than T4 DNA Ligase, as well as a preference for A/T overhangs (as opposed to C/G overhang pairs).

Specifications

Packaging Vial
Enzyme DNA Ligase
Sensitive to Heat Inactivation Yes
Product Type T3 DNA Ligase
Research Category Traditional Cloning, Next-generation Sequencing, DNA Repair
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.