Learn More
Simport™ Scientific T310 Series Cryovials
Catalog No. 22039100 Shop All Simport Scientific Products
Change view
Click to view available options
Volume (Metric):
3 mL
4 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-039-100 10 mL
10-269-88F 4 mL
10-269-88D 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-039-100 Supplier Simport™ Scientific Supplier No. T31010A
Add to Cart
Add to Cart
This item is not returnable. View return policy
T310 Series Cryovials are designed for storing biological material, human or animal cells, these cryovials are compatible with most storage systems.
  • Cap features a long skirt for easy one-hand aseptic methods and an exclusive silicone seal fitted inside the cap
  • Super-fast thread design allows removal or sealing with a mere 1 1/4 turn
  • Inside thread design that will not contribute to possible contamination
  • Tubes are provided with a white marking area for sample identification
  • Sterilized by gamma radiation
  • Autoclavable
  • Polypropylene construction
Specifications
Material Polypropylene
Sterility Sterile
Volume (Metric) 10 mL
For Use With (Application) For storing biological material, human or animal cells, these cryovials are compatible with most storage systems
Thread Style External
RNase Free Yes
Dnase Free Yes
Closure Type Screw Cap
Quantity 500/Cs.
Marking Spot Yes
Autoclavable Autoclavable
Product Type Cryogenic Vial