Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences T4 DNA Ligase
SCI_ED

Catalog No. S03116
Change view
Click to view available options
Quantity:
20,000 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03116 20,000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03116 Supplier G-Biosciences Supplier No. 7862001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T4 DNA Ligase is used for cloning of restriction enzyme generated DNA fragments and PCR products. It is also used for next-generation library preparation, joining linkers and adapters to cohesive or blunt-ended DNA, self-circularization of linear DNA.

  • T4 DNA Ligase catalyzes the formation of a phosphodiester bond between neighboring 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.
  • It can be used to join DNA fragments with both sticky ends and blunt ends, and to repair nicks in double-stranded DNA .

Specifications

Enzyme DNA Ligase
Sensitive to Heat Inactivation Yes
Quantity 20,000 Units
Product Type DNA Ligase
Research Category Genome Editing, Traditional Cloning, Scarless Cloning, Next-generation Sequencing, DNA Repair
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.