Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ T4 DNA Ligase (5 U/μL)

Catalog No. 15224041
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-224-041 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-224-041 Supplier Invitrogen™ Supplier No. 15224041
Only null left
Add to Cart
Edge
Add to Cart

Includes

T4 DNA Ligase is supplied with a vial of 5X reaction buffer [250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (w/v) polyethylene glycol-8000].

Catalyzes the formation of phosphodiester bonds in the presence of ATP between double-stranded DNAs with 3' hydroxyl and 5' phosphate termini

The unique T4 DNA Ligase buffer optimizes ligation, which can be performed in 5 minutes. Single-stranded nucleic acids are not substrates for this enzyme.

Unit Definition: One unit catalyzes the exchange of 1nmol 32P-labeled pyrophosphate into ATP in 20 min. at 37°C. (One unit is equal to approximately 300 cohesive-end ligation units.)

Unit Reaction Conditions: 66mM Tris-HCl (pH 7.6), 6.6mM MgCl2 , 10mM DTT, 66μM ATP, 3.3μM 32P-labeled pyrophosphate, and enzyme in 0.1mL for 20 min. at 37°C.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage T4 DNA Ligase is supplied with a vial of 5X reaction buffer [250 mM Tris-HCl (pH 7.6), 50 mM MgCl2 , 5 mM ATP, 5 mM DTT, 25% (w/v) polyethylene glycol-8000]. Store at -20°C.
Shipping Condition Dry Ice
Enzyme Ligase
Compatible Buffer 5X Reaction Buffer
Quantity 250 U
Product Type T4 DNA Ligase
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.