Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences T4 DNA Polymerase
SCI_ED

Catalog No. S03112
Change view
Click to view available options
No. of Reactions:
200
750
1500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03112 200
S03108 1500
S03110 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S03112 Supplier G-Biosciences Supplier No. 7862004
Add to Cart
Add to Cart

T4 DNA Polymerase is used for 3'-overhang removal to form blunt ends, 5'-overhang fill-in to form blunt ends, probe labeling and DNA library preparation for Next-generation sequencing.

  • T4 DNA Polymerase catalyzes the synthesis of DNA in the 5' - 3' direction with the presence of template and primer.
  • It has both DNA-dependent DNA polymerase activity and 3' - 5' exonuclease activity which make the enzyme useful for several molecular biology applications.

Specifications

Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance Low
PCR Method Standard PCR, End-point PCR, qPCR, Multiplex qPCR
Reaction Speed Standard
Label or Dye Biotin and Other Hapten, No ROX, Biotin, Unlabeled, Unconjugated
Product Type DNA Polymerase
For Use With (Application) Pathogen Detection, Non-Radioactive Labeling, DNA Sequencing, Next-Generation Sequencing (NGS), Cloning
No. of Reactions 200
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.