Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T4 RNA ligase I, 5000 U, G-Biosciences
SCI_ED

Catalog No. S001142
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001142 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001142 Supplier G-Biosciences Supplier No. 7862277
Add to Cart
Add to Cart

T4 RNA Ligase I (gp63) catalyzes the ATP-dependent ligation of single-stranded nucleic acids by joining a 5′ phosphate to a 3′ hydroxyl. It is most efficient at ligating the 3′-OH of RNA to the 5′-phosphate of RNA or DNA in single-stranded contexts.

  • T4 RNA Ligase I is commonly used for 3' adapter ligation in small RNA library prep, RNA circularization, and labeling of RNA or DNA with linkers.

Specifications

Packaging Vial
Enzyme RNA Ligase
Sensitive to Heat Inactivation Yes
Product Type T4 RNA Ligase I
Research Category RNA Research, Next-generation Sequencing
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.