Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T4 RNA ligase II, 750 U, G-Biosciences
SCI_ED

Catalog No. S001108
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001108 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001108 Supplier G-Biosciences Supplier No. 7862278
Add to Cart
Add to Cart

T4 RNA Ligase II (gp24.1) repairs nicks in dsRNA and can also be used to ligate the 3' terminal OH of RNA to the 5'-terminal phosphate of DNA in a double-stranded structure.

  • T4 RNA Ligase 2 will also join the 3'-end of RNA or DNA to a 5'-pDNA when annealed to an RNA complement, and, with reduced efficiency, the 3'-end of RNA to a 5'-pRNA when annealed to a DNA complement.

Specifications

Packaging Vial
Enzyme RNA Ligase II
Sensitive to Heat Inactivation Yes
Product Type T4 RNA Ligase 2
Research Category RNA Research, DNA Repair
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.