Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T4 UvsX DNA Recombinase, Lyophilized, G-Biosciences
SCI_ED

Catalog No. S000285
Change view
Click to view available options
Quantity:
100 μg
1000 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000285 100 μg
S000268 500 μg
S000272 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S000285 Supplier G-Biosciences Supplier No. 7862132
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes.

The protein forms a right-handed nucleoprotein complex on single ssDNA called the presynaptic filament that can search for homology in duplex DNA and pair the recombining DNA molecules to form a DNA joint molecule. This process is aided by ssDNA binding proteins and recombination mediators. The filament is then elongated by DNA polymerase with the newly synthesized strand displacing the old strand. The newly synthesized duplex DNA can then act as a template for the next cycle to achieve exponential amplification of dsDNA

Specifications

Detection Method Gel Electrophoresis
Form Lyophilized
GC-Rich PCR Performance High
PCR Method End-point PCR
Reaction Speed 20 min.
Label or Dye Biotin, FAM, TAMRA
Product Type Uvsx Recombinase
Quantity 100 μg
For Use With (Application) Microbial Detection, Pathogen Detection, Isothermal Amplification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.