Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T7 DNA Ligase, 75000 U, G-Biosciences
SCI_ED

Catalog No. S001162
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001162 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001162 Supplier G-Biosciences Supplier No. 7862276
Add to Cart
Add to Cart

T7 DNA Ligase is an ATP-dependent enzyme from bacteriophage T7 that catalyzes phosphodiester bond formation between 5′ phosphate and 3′ hydroxyl termini of duplex DNA.

  • It efficiently ligates nicks and cohesive ends but lacks blunt-end ligation activity.

Specifications

Packaging Vial
Enzyme T7 DNA Ligase
Sensitive to Heat Inactivation Yes
Product Type T7 DNA Ligase
Research Category Traditional Cloning, Next-generation Sequencing
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.