Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T7 Exonuclease, 2500 U, G-Biosciences
SCI_ED

Catalog No. S001160
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001160 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001160 Supplier G-Biosciences Supplier No. 7862271
Add to Cart
Add to Cart

Gene 6 protein of bacteriophage T7 is a dsDNA 5' to 3' exonuclease which also exhibits flap endonuclease activity.

  • T7 Exonuclease also degrades RNA and DNA from RNA:DNA hybrids in the 5'→3' direction, however it is unable to degrade either double-stranded or single-stranded RNA.

Specifications

Packaging Vial
Enzyme T7 Exonuclease
Sensitive to Heat Inactivation Yes
Research Category RNA Research, Traditional Cloning
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.