Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAAR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545006 Supplier Thermo Scientific Supplier No. PA545006
Only null left
Add to Cart
Add to Cart

TAAR5 Polyclonal Antibody

TAAR5 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen TAAR5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Taar5
Gene Accession No. O14804, Q5QD14
Gene Alias Gm227; hTaar5; MGC138414; MGC138416; mTaar5; PNR; Putative neurotransmitter receptor; RP11-295F4.5; TAAR5; TaR-5; trace amine associated receptor 5; trace amine receptor 5; trace amine-associated receptor 5; Trimethylamine receptor
Gene Symbols Taar5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TAAR5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215854, 9038
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.