Learn More
Bachem Tachyplesin I Trifluoroacetate
SDP
Supplier:  Bachem 4030734.0500
Encompass_Preferred
Tachyplesin I Trifluoroacetate, H-Lys-Trp-Cys-Phe-Arg-Val-Cys-Tyr-Arg-Gly-Ile-Cys-Tyr-Arg-Arg-Cys-Arg-NH2, 118231-04-2, C99H151N35O19S4, Mr 2263.78, 0.5mg. Tachyplesin I is an antimicrobial peptide originally detected in the extract of hemocytes from the Japanese horseshoe crab. It inhibits the growth of both Gramnegative and -positive bacteria and forms a complex with bacterial lipopolysaccharides. These results suggest that this peptide belongs to the family of arthropodous peptide antibiotics. Furthermore this peptide has been shown to have antiviral activity against vesicular stomatitis virus, influenza A virus and HIV-1.
Catalog No. 50-259-475
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.