Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TACR1 Monoclonal Antibody (ZN003)
GREENER_CHOICE

Catalog No. 396100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-610-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-610-0 Supplier Invitrogen™ Supplier No. 396100
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

TACR1 Monoclonal Antibody for Western Blot, ICC/IF, ELISA

Tachykinin Receptor 1, a member of the Tachykinin Receptor subfamily, is involved in the mediation of phosphatidylinositol metabolism by the tachykinin substance P, also referred to as neurokinin 1. It is associated with the management of depression, anxiety, and stress. It is also involved in regulating behavioral responses, neuronal survival, and cardiovascular and respiratory functions. The tachykinin receptor 1 is implicated in the progression of rheumatoid arthritis and osteoarthritis. Two variants of the tachykinin receptor 1 are produced by alternative splicing. Tachykinin receptor 1 has been reported primarily in brain, and in blood lymphocytes, nose, small intestine, and stomach. ESTs have been isolated from brain and spleen libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACR1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ZN003
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Tacr1
Gene Accession No. P14600, P25103, P30548
Gene Alias neurokinin 1 receptor; neurokinin receptor 1; neurokinin receptor subtype NK1; neurokinin-1 receptor; Nk1 receptor; NK-1 receptor; Nk1r; NK-1R; NKIR; SPR; substance p receptor; substance-P receptor; TAC1R; tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor); TACR1; truncated neurokinin-1 receptor
Gene Symbols Tacr1
Host Species Mouse
Immunogen Synthetic peptide derived from an internal sequence of the human, mouse, and rat NK1 receptor proteins.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21336, 24807, 6869
Target Species Human, Mouse, Rat, Insect
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.