Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TACR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APS301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APS301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APS301 Supplier Invitrogen™ Supplier No. PA3301
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA3-301 detects neurokinin-1 receptor in human samples. PA3-301 has been successfully used in Western blot and immunohistochemistry (paraffin) procedures. The PA3-301 immunogen is a synthetic peptide corresponding to residues S(387) S R S D S K T M T E S F S F S S N V L S(407) of human NK1 receptor protein.

Tachykinin Receptor 1, a member of the Tachykinin Receptor subfamily, is involved in the mediation of phosphatidylinositol metabolism by the tachykinin substance P, also referred to as neurokinin 1. It is associated with the management of depression, anxiety, and stress. It is also involved in regulating behavioral responses, neuronal survival, and cardiovascular and respiratory functions. The tachykinin receptor 1 is implicated in the progression of rheumatoid arthritis and osteoarthritis. Two variants of the tachykinin receptor 1 are produced by alternative splicing. Tachykinin receptor 1 has been reported primarily in brain, and in blood lymphocytes, nose, small intestine, and stomach. ESTs have been isolated from brain and spleen libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene Tacr1
Gene Accession No. P25103
Gene Alias neurokinin 1 receptor; neurokinin receptor 1; neurokinin receptor subtype NK1; neurokinin-1 receptor; Nk1 receptor; NK-1 receptor; Nk1r; NK-1R; NKIR; SPR; substance p receptor; substance-P receptor; TAC1R; tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor); TACR1; truncated neurokinin-1 receptor
Gene Symbols Tacr1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues S(387) S R S D S K T M T E S F S F S S N V L S(407) of human NK1 receptor protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6869
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.