Learn More
Medchemexpress LLC Tacrolimus 8-epimer | 129212-35-7 | MFCD27967171 | 10mg
Supplier: Medchemexpress LLC HYZ875410MG
Tacrolimus anhydrous 8-epimer (Tacrolimus EP impurity D) is a l-pipecolic acid macrolide lactone an important immunosuppressive agent that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines especially IL-2[1]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.