Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tacrolimus, 99+%

Catalog No. p-7082362
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63571ME 500 mg
AAJ63571MF 50 mg
AAJ63571MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63571ME Supplier Thermo Scientific Chemicals Supplier No. J63571ME
Only null left
Add to Cart

Tacrolimus, CAS # 104987-11-3, also known as FK-506 or fujimycin, is a macrolide compound that exhibits immunosuppressant, neuroprotective, and neuroregenerative activities and can inhibit T-cell proliferation and activation. | CAS: 104987-11-3 | C44H69NO12

Interacts with the immunophilins cyclophilin and FKBP-12.Tacrolimus is used as an immunosuppressive drug that blocks T cell proliferation in vitro. Further, it is used as a research tool in chemically induced dimerization applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Tacrolimus is a macrolide isolated from Streptomyces tsukubaensis, that shows immunosuppressant, neuroprotective, and neuroregenerative activities, having the ability to block T-cell proliferation and activation in response to antigenic or mitogenic stimulation

• Tacrolimus interacts with the immunophilins cyclophilin and FKBP-12. Tacrolimus binds to the FKBP-12 protein (FK506 binding protein) and forms a complex with calcium-dependent proteins. This FKBP12-FK506 complex inhibits calcineurin phosphatase activity, resulting in decreased cytokine production, inhibition of T-lymphocyte signal transduction

• Tacrolimus also inhibits the transcription for genes that encode IL-3, IL-4, IL-5, GM-CSF, and TNF, all of which are involved in the early stages of T-cell activation

• This compound can also inhibit the release of pre-formed mediators from skin mast cells and basophils and downregulate the expression of FceRI on Langerhans cells

Application

• Tacrolimus has been studied in vitro to understand its impact on neutrophil production and/or survival • It is applied to suppress the inflammation in ulcerative cells

• It is used as a research tool in chemically induced dimerization applications in biology

• It has been used in in vitro studies to investigate its effect on human monocyte-derived dentritic cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104987-11-3
Molecular Formula C44H69NO12
Synonym FK-506; Fugimycin

Specifications

Quantity 500 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Interacts with the immunophilins cyclophilin and FKBP-12
Formula Weight 804.02g/mol
Melting Point 120°C to 125°C
Solubility Freely soluble in DMSO or ethanol; Poorly soluble in water
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%
At what temperature should I store Tacrolimus, 99+%?

Tacrolimus should be stored in the freezer at -20 degrees Celsius.

What is the solubility of Tacrolimus, 99+%?

Tacrolimus may be dissolved in DMSO (50 mg/mL) or ethanol (50 mg/mL). This compound is very poorly soluble in water.

Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.