Learn More
Medchemexpress LLC Tacrolimus (FK506) | 104987-11-3 | MFCD00869853 | 99.6% | 803.48 Da | C44H69NO12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13756R5MG
Encompass_Preferred
Tacrolimus (FK506) analytical standard intended for research and analytical applications. Tacrolimus is a macrolide immunosuppressant that binds FK506 binding protein and inhibits calcineurin, affecting T-lymphocyte signal transduction. This reference material is supplied for assay calibration, method development, and quality control in analytical workflows.
  • High-purity reference material (99.6%).
  • Intended for analytical and research use, including assay calibration and method development.
  • Available in small mass quantities suitable for analytical workflows (e.g., 5 mg).
  • Provides a stable, characterized standard for quality control.
  • Supplied with documentation for purity and identity.
Catalog No. 50-002-21966