Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 10X TdT Stop Buffer
SDP

Catalog No. 48103003
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-103-003 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-103-003 Supplier R&D Systems™ Supplier No. 48103003

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ TACS 2 TdT Stop Buffer is provided with the TACS TdT kits to stop the activity of the TdT enzyme in TUNEL labeling assays.

  • Uncontrolled product according to WHMIS classification criteria
  • All of the ingredients in this product are listed on the United States TSCA (Toxic Substances Control Act) inventory, the Domestic Substances List (DSL) and/or the Non-Domestic Substances List (NDSL)

Specifications

Quantity 100 mL
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.