Learn More
Novus Biologicals™ TACS-XL™ In Situ Apoptosis Kit [TACS Blue Label]
Description
- TUNEL assay
- Incorporates bromodeoxyuridine (BrdU) at 3’ OH ends of the DNA fragments that form during apoptosis
- Uses biotin conjugated anti-BrdU antibody, streptavidin-horseradish peroxidase
Detection: Biotinylated dNTP/Streptavidin-HRP/Blue Label
Testing Format: Light Microscopy
Sample Type: Fixed cells, Embedded tissues, Frozen tissues
Specifications
Specifications
| Quantity | 30 Tests |
| Product Type | Apoptosis detection kit |