Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Tacta™ Adapter

Catalog No. 14577819 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
6-place Carousel Stand
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-577-819 6-place Carousel Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-577-819 Supplier Sartorius Supplier No. LH727650
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tacta™ Adapter for Mechanical Carousel Stand.

  • The Tacta™ Adapter for the 6-Place Carousel Stand is designed for use with all Sartorius Tacta mechanical pipettes.
  • It is recommended for the safe storage of Tacta pipettes when not in use to avoid contamination and/or damage to the pipettes.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Pipettes
Material Polypropylene
Product Line Tacta
Sterility Non-sterile
Type Adapter
For Use With (Application) Safe Storage of Pipettes
For Use With (Equipment) 6-place Carousel Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.