Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Tacta™ Pipette Holder

Catalog No. 14577820 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
0.384 dL3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-577-820 0.384 dL3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-577-820 Supplier Sartorius Supplier No. LH727640
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tacta™ Pipette Holder for one Tacta™ pipette.

  • Pipette holder for one pipette - The pipette holder can be mounted on a table edge or on a shelf.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
Material Polypropylene
Product Line Tacta
Sterility Non-sterile
Type Holder
Volume (Metric) 0.384 dL3
For Use With (Application) Can be mounted on a table edge or on a shelf
For Use With (Equipment) Tacta Pipette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.