Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tadocizumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA541731
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Tadocizumab is a biosimilar that targets ITGA2B & ITGB3. CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GpiIb alpha (heavy chain; 120 kDa) and GpiIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
Specifications
| Tadocizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| C4G1; YM-337 | |
| Human ITGA2B_ITGB3. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 2.15 mg/mL | |
| PBS with no preservative; pH 7.5 | |
| Human | |
| Protein L, Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 F(ab) κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction