Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TAE (50X), TRIS + acetate + EDTA

Catalog No. AAJ63931K3
Change view
Click to view available options
Quantity:
1 L
2 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63931K3 2 L
AAJ63931K2 1 L
AAJ63931K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63931K3 Supplier Thermo Scientific Chemicals Supplier No. J63931K3
Add to Cart

TAE (50X), TRIS + acetate + EDTA is a buffer composed of Tris, acetate, and EDTA components and is used in electrophoresis of nucleic acids in agarose and polyacrylamide gels.

TAE (50X), TRIS + acetate + EDTA is used in electrophoresis of nucleic acids in agarose and polyacrylamide gel. It acts as a running buffer and as a gel preparation buffer. It is also useful for nucleic acid separations viz. DNA and RNA. Further, it is used for non-denaturing RNA agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• TAE (50X), TRIS + acetate + EDTA allows nucleic acid separation

Application

• This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
• It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Description Colorless
Concentration 50X
Physical Form Liquid
Quantity 2 L

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.