Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Grade 10X TAE Buffer, Promega

Catalog No. PRV4271 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-V4271 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-V4271 Supplier Promega Corporation Supplier No. V4271
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Commonly used buffer for agarose DNA electrophoresis.

  • Quality tested and certified free of DNase, RNase and protease activity

Chemical Identifiers

CAS 77-86-1 , 7732-18-5
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

For Use With (Application) Agarose Gel Electrophoresis, Polyacrylamide Gel Electrophoresis
pH 8.2 to 8.4
Physical Form Liquid
Quantity 1000 mL
Solution Type Buffer
Grade Electrophoresis
Chemical Name or Material TAE Buffer 10X
Recommended Storage 15 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.