Learn More
Description
- Quality tested and certified free of DNase, RNase and protease activity
Specifications
Specifications
| Chemical Name or Material | TAE Buffer 40X |
| CAS | 7732-18-5 |
| For Use With (Application) | Electrophoresis |
| pH | 8.2 to 8.4 |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Solution Type | Buffer |
| Grade | Electrophoresis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.