Learn More
TAF1A Polyclonal Antibody

Description
TAF1A Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | TAF1A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TAF1A |
| Gene Accession No. | P97357, Q15573, Q3B7U2 |
| Gene Alias | MGC:17061; mTAFI48; RAFI48; RNA polymerase I-specific TBP-associated factor 48 kDa; similar to TBP-associated factor 1A isoform 1; SL1; SL1, 48kD subunit; Taf1a; TAFI48; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, A; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa; TATA box binding protein associated factor 1A; TATA box binding protein-associated factor TAF1A; TATA box-binding protein-associated factor 1A; TATA box-binding protein-associated factor RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I, A; TBP-associated factor 1A; Transcription factor SL1; transcription initiation factor SL1/TIF-IB subunit A |
| Gene Symbols | TAF1A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.