Learn More
Aviva Systems Biology Corp TAF1B Antibody - C-terminal region (ARP37289_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37289_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse TAF1B. Alternate names: TAFI68, Tafi86, 4930408G01Rik, A230108M10RikApplicationWB. Species reactivity: Guinea Pig, Mouse, Rat
Catalog No. 50-014-56346