Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568845 Supplier Invitrogen™ Supplier No. PA568845
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 79%.

Component of RNA polymerase I core factor complex that acts as a GTF2B/TFIIB-like factor and plays a key role in multiple steps during trancription initiation such as preinitiation complex (PIC) assembly and postpolymerase recruitment events in polymerase I (Pol I) transcription. Binds rDNA promoters and plays a role in Pol I recruitment as a component of the SL1/TIF-IB complex and, possibly, directly through its interaction with RRN3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TAF1B
Gene Accession No. P97358
Gene Alias 4930408G01Rik; A230108M10Rik; MGC:9349; p63; RAF1B; RAFI63; RNA polymerase I-specific TBP-associated factor 63 kDa; RNA polymerase I-specific TBP-associated factor 68 kDa; SL1; SL1, 63kD subunit; Taf1b; TAFI63; TAFI68; Tafi86; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, B; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa; TATA box binding protein associated factor 1B; TATA box-binding protein-associated factor 1B; TATA box-binding protein-associated factor RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I, B; TBP-associated factor 1B; TBP-associated factor RNA polymerase I 68; transcription factor TAFI68; transcription initiation factor SL1/TIF-IB subunit B
Gene Symbols TAF1B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse TAF1B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21340
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.