Learn More
Medchemexpress LLC Tak-418 hydrochloride | 1818252-53-7 | 99.8% | C17H25ClN2O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13883010MG
Encompass_Preferred
TAK-418 is an orally active, CNS-penetrant inhibitor of lysine-specific demethylase 1 (LSD1/KDM1A) developed as a clinical candidate for neurodevelopmental and other central nervous system disorders. It is supplied as a research compound for in vitro and in vivo pharmacology, pharmacokinetics, and translational studies, with reported formula C17H25ClN2O2S and CAS 1818252-53-7.
  • Selective LSD1 inhibition with reported low-nanomolar potency.
  • Orally bioavailable and brain-penetrant for CNS studies.
  • Suitable for in vitro and in vivo pharmacology.
  • Available in small-mg research pack sizes for preclinical work.
  • High reported purity for reproducible experimental results.
  • Storage guidance supports long-term sample integrity.
Catalog No. 50-002-61840
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.