Learn More
TARGETMOL CHEMICALS INC TAK-418 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T39252
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TAK-418 is a selective and orally active inhibitor of LSD1/KDM1A enzyme with an IC50 of 2.9 nM. TAK-418 unlocks abnormal epigenetic mechanisms and improves autism symptoms in models of neurodevelopmental disorders. purity: 98%
Catalog No. 50-258-0061
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.