Learn More
Apexbio Technology LLC Fluvoxamine maleate 61718-82-9 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC A25535.1
Fluvoxamine maleate (CAS 61718-82-9) is a selective serotonin reuptake inhibitor that acts by inhibiting the human serotonin (5-HT) transporter This action leads to increased extracellular serotonin concentrations Fluvoxamine maleate exhibits high binding affinity for the 5-HT transporter with a reported K sub i /sub of 1 6 nmol/l It is widely utilized in biomedical research to investigate serotonergic signaling pathways and to model mechanisms underlying antidepressant activity Fluvoxamine maleate is also available as part of the Serotonin Uptake Inhibitor Tocriset for experimental applications
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.